Assay Details
| Gene Symbol: LOC4344427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795268.2 | XP_015650754.1 | 1 - 2 | 595 | 84 | |
| XM_015795269.2 | XP_015650755.1 | 1 - 2 | 595 | 84 | ||
| XM_015795270.2 | XP_015650756.1 | 1 - 2 | 595 | 84 | ||
| XM_015795271.2 | XP_015650757.1 | 2 - 3 | 496 | 84 | ||
| GenBank mRNA | AK067246.1 | - | 2 - 3 | 307 | 84 | |
More Information
Set Membership: |
|

LOC4344427
Genomic Map