Assay Details
| Gene Symbol: LOC4344427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795268.2 | XP_015650754.1 | 5 - 6 | 979 | 139 | |
| XM_015795269.2 | XP_015650755.1 | 5 - 6 | 979 | 139 | ||
| XM_015795270.2 | XP_015650756.1 | 5 - 6 | 979 | 139 | ||
| XM_015795271.2 | XP_015650757.1 | 6 - 7 | 880 | 139 | ||
| GenBank mRNA | AK067246.1 | - | 6 - 7 | 691 | 139 | |
More Information
Set Membership: |
|

LOC4344427
Genomic Map