Assay Details
| Gene Symbol: LOC4344427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795268.2 | XP_015650754.1 | 8 - 9 | 1341 | 61 | |
| XM_015795269.2 | XP_015650755.1 | 8 - 9 | 1341 | 61 | ||
| XM_015795270.2 | XP_015650756.1 | 8 - 9 | 1341 | 61 | ||
| XM_015795271.2 | XP_015650757.1 | 9 - 10 | 1242 | 61 | ||
| GenBank mRNA | AK061938.1 | - | 1 - 2 | 184 | 61 | |
| AK067246.1 | - | 9 - 10 | 1053 | 61 | ||
More Information
Set Membership: |
|

LOC4344427
Genomic Map