Assay Details
| Gene Symbol: LOC4344608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795380.2 | XP_015650866.1 | 9 - 10 | 1505 | 117 | |
| GenBank mRNA | AK064276.1 | - | 9 - 10 | 1289 | 117 | |
| AK072961.1 | - | 9 - 10 | 1300 | 117 | ||
More Information
Set Membership: |
|

LOC4344608
Genomic Map