Assay Details
| Gene Symbol: LOC4344614 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793053.2 | XP_015648539.1 | 2 - 3 | 964 | 80 | |
| GenBank mRNA | AB101650.1 | - | 1 - 2 | 171 | 80 | |
| AK318563.1 | - | 2 - 3 | 920 | 80 | ||
More Information
Set Membership: |
|

LOC4344614
Genomic Map