Assay Details
| Gene Symbol: LOC4344621 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794401.1 | XP_015649887.1 | 19 - 20 | 1429 | 82 | |
| GenBank mRNA | AK065561.1 | - | 19 - 20 | 1395 | 82 | |
| AK101582.1 | - | 19 - 20 | 1395 | 82 | ||
More Information
Set Membership: |
|

LOC4344621
Genomic Map