Assay Details
| Gene Symbol: LOC4344685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794626.2 | XP_015650112.1 | 7 - 8 | 2082 | 138 | |
| GenBank mRNA | AK070325.1 | - | 7 - 8 | 2083 | 138 | |
| AK099134.1 | - | 7 - 8 | 2062 | 138 | ||
More Information
Set Membership: |
|

LOC4344685
Genomic Map