Assay Details
| Gene Symbol: LOC4344785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794372.1 | XP_015649858.1 | 4 - 5 | 715 | 129 | |
| XM_015794373.2 | XP_015649859.1 | 4 - 5 | 682 | 129 | ||
| GenBank mRNA | AB188247.1 | - | 4 - 5 | 666 | 129 | |
| AK103038.1 | - | 4 - 5 | 721 | 129 | ||
More Information
Set Membership: |
|

LOC4344785
Genomic Map