Assay Details
| Gene Symbol: LOC4344785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794372.1 | XP_015649858.1 | 6 - 7 | 855 | 130 | |
| XM_015794373.2 | XP_015649859.1 | 6 - 7 | 822 | 130 | ||
| GenBank mRNA | AB188247.1 | - | 6 - 7 | 806 | 130 | |
| AK103038.1 | - | 6 - 7 | 861 | 130 | ||
More Information
Set Membership: |
|

LOC4344785
Genomic Map