Assay Details
| Gene Symbol: LOC4344798 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795639.2 | XP_015651125.1 | 1 - 2 | 188 | 58 | |
| GenBank mRNA | AK111670.1 | - | 1 - 2 | 193 | 58 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015795639.2 transcripts.
Set Membership: |
|

LOC4344798
Genomic Map