Assay Details
| Gene Symbol: LOC4344798 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795639.2 | XP_015651125.1 | 7 - 8 | 830 | 113 | |
| XM_015795640.2 | XP_015651126.1 | 7 - 8 | 824 | 113 | ||
| GenBank mRNA | AK111670.1 | - | 7 - 8 | 835 | 113 | |
More Information
Set Membership: |
|

LOC4344798
Genomic Map