Assay Details
| Gene Symbol: LOC4344798 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795639.2 | XP_015651125.1 | 9 - 10 | 1032 | 90 | |
| XM_015795640.2 | XP_015651126.1 | 9 - 10 | 1026 | 90 | ||
| GenBank mRNA | AK111670.1 | - | 9 - 10 | 1037 | 90 | |
More Information
Set Membership: |
|

LOC4344798
Genomic Map