Assay Details
| Gene Symbol: LOC4344801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794248.2 | XP_015649734.1 | 1 - 2 | 194 | 86 | |
| XM_015794249.2 | XP_015649735.1 | 1 - 2 | 222 | 86 | ||
| GenBank mRNA | AK063414.1 | - | 1 - 2 | 271 | 86 | |
| AK101719.1 | - | 1 - 2 | 228 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015794248.2, XM_015794249.2 transcripts.
Set Membership: |
|

LOC4344801
Genomic Map