Assay Details
| Gene Symbol: LOC4344818 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792921.2 | XP_015648407.2 | 3 - 4 | 415 | 62 | |
| GenBank mRNA | AK063927.1 | - | 3 - 4 | 383 | 62 | |
| AK103587.1 | - | 3 - 4 | 421 | 62 | ||
More Information
Set Membership: |
|

LOC4344818
Genomic Map