Assay Details
| Gene Symbol: LOC4344818 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792921.2 | XP_015648407.2 | 8 - 9 | 840 | 137 | |
| GenBank mRNA | AK063927.1 | - | 8 - 9 | 808 | 137 | |
| AK103587.1 | - | 8 - 9 | 846 | 137 | ||
More Information
Set Membership: |
|

LOC4344818
Genomic Map