Assay Details
| Gene Symbol: LOC4345048 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792973.2 | XP_015648459.1 | 5 - 6 | 676 | 116 | |
| GenBank mRNA | AB110183.1 | - | 5 - 6 | 658 | 116 | |
| AK101443.1 | - | 5 - 6 | 667 | 116 | ||
More Information
Set Membership: |
|

LOC4345048
Genomic Map