Assay Details
| Gene Symbol: LOC4345257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795548.1 | XP_015651034.1 | 6 - 7 | 930 | 69 | |
| GenBank mRNA | AK243589.1 | - | 5 - 6 | 1023 | 69 | |
More Information
Set Membership: |
|

LOC4345257
Genomic Map