Assay Details
| Gene Symbol: LOC4345288 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795587.2 | XP_015651073.1 | 1 - 2 | 392 | 73 | |
| XM_015795588.2 | XP_015651074.1 | 1 - 2 | 392 | 73 | ||
| GenBank mRNA | AK102227.1 | - | 1 - 2 | 393 | 73 | |
| AK105392.1 | - | 1 - 2 | 350 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015795587.2, XM_015795588.2 transcripts.
Set Membership: |
|

LOC4345288
Genomic Map