Assay Details
| Gene Symbol: LOC4345327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795449.2 | XP_015650935.1 | 1 - 2 | 362 | 82 | |
| GenBank mRNA | AK072532.1 | - | 1 - 2 | 265 | 82 | |
| AK120904.1 | - | 1 - 2 | 262 | 82 | ||
| Gene Symbol: LOC4328287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK287728.1 | - | 217 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015795449.2 transcripts.
Set Membership: |
|

LOC4345327
Genomic Map