Assay Details
| Gene Symbol: LOC4345327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795449.2 | XP_015650935.1 | 3 - 4 | 796 | 74 | |
| GenBank mRNA | AK072532.1 | - | 3 - 4 | 699 | 74 | |
| AK120904.1 | - | 3 - 4 | 696 | 74 | ||
More Information
Set Membership: |
|

LOC4345327
Genomic Map