Assay Details
| Gene Symbol: LOC4345415 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795104.1 | XP_015650590.1 | 1 - 2 | 477 | 126 | |
| GenBank mRNA | AB096863.1 | - | 1 - 2 | 437 | 126 | |
| AK066269.1 | - | 1 - 2 | 462 | 126 | ||
More Information
Set Membership: |
|

LOC4345415
Genomic Map