Assay Details
| Gene Symbol: LOC107281818 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792815.2 | XP_015648301.1 | 2 - 3 | 2572 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107281818| Gene Symbol: LOC107281818 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792815.2 | XP_015648301.1 | 2 - 3 | 2572 | 75 | |
Set Membership: |
|