Assay Details
| Gene Symbol: LOC4346285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792790.2 | XP_015648276.1 | 1 - 2 | 322 | 57 | |
| GenBank mRNA | AK121222.1 | - | 1 - 2 | 225 | 57 | |
| BT129121.1 | - | 1 - 2 | 328 | 57 | ||
More Information
Set Membership: |
|

LOC4346285
Genomic Map