Assay Details
| Gene Symbol: LOC4346318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794261.2 | XP_015649747.1 | 3 - 4 | 701 | 118 | |
| GenBank mRNA | AK058527.1 | - | 3 - 4 | 669 | 118 | |
| AK102083.1 | - | 3 - 4 | 703 | 118 | ||
| AK103178.1 | - | 3 - 4 | 708 | 118 | ||
More Information
Set Membership: |
|

LOC4346318
Genomic Map