Assay Details
| Gene Symbol: LOC4346318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794261.2 | XP_015649747.1 | 8 - 9 | 1286 | 98 | |
| GenBank mRNA | AK058527.1 | - | 8 - 9 | 1254 | 98 | |
| AK102083.1 | - | 8 - 9 | 1288 | 98 | ||
| AK103178.1 | - | 8 - 9 | 1293 | 98 | ||
More Information
Set Membership: |
|

LOC4346318
Genomic Map