Assay Details
| Gene Symbol: LOC4346318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794261.2 | XP_015649747.1 | 9 - 10 | 1419 | 125 | |
| GenBank mRNA | AK058527.1 | - | 9 - 10 | 1387 | 125 | |
| AK102083.1 | - | 9 - 10 | 1421 | 125 | ||
| AK103178.1 | - | 9 - 10 | 1426 | 125 | ||
More Information
Set Membership: |
|

LOC4346318
Genomic Map