Assay Details
| Gene Symbol: LOC4347429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015796002.2 | XP_015651488.1 | 1 - 2 | 275 | 140 | |
| XM_015796003.2 | XP_015651489.1 | 1 - 2 | 275 | 140 | ||
| XM_015796004.2 | XP_015651490.1 | 1 - 2 | 275 | 140 | ||
| XM_015796005.2 | XP_015651491.1 | 1 - 2 | 275 | 140 | ||
| XM_015796007.2 | XP_015651493.1 | 1 - 2 | 275 | 140 | ||
| GenBank mRNA | AK070222.1 | - | 1 - 2 | 282 | 140 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015796002.2, XM_015796003.2, XM_015796004.2, XM_015796005.2, XM_015796007.2 transcripts.
Set Membership: |
|

LOC4347429
Genomic Map