Assay Details
| Gene Symbol: LOC4347429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015796002.2 | XP_015651488.1 | 2 - 3 | 800 | 90 | |
| XM_015796003.2 | XP_015651489.1 | 2 - 3 | 800 | 90 | ||
| XM_015796004.2 | XP_015651490.1 | 2 - 3 | 800 | 90 | ||
| XM_015796005.2 | XP_015651491.1 | 2 - 3 | 800 | 90 | ||
| XM_015796007.2 | XP_015651493.1 | 2 - 3 | 800 | 90 | ||
| XM_015796008.2 | XP_015651494.1 | 3 - 4 | 715 | 90 | ||
| XM_015796009.2 | XP_015651495.1 | 3 - 4 | 650 | 90 | ||
| GenBank mRNA | AK070222.1 | - | 2 - 3 | 807 | 90 | |
| EU605826.1 | - | 1 - 2 | 487 | 90 | ||
More Information
Set Membership: |
|

LOC4347429
Genomic Map