Assay Details
| Gene Symbol: LOC4347429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015796002.2 | XP_015651488.1 | 5 - 6 | 1062 | 78 | |
| XM_015796003.2 | XP_015651489.1 | 5 - 6 | 1062 | 78 | ||
| XM_015796004.2 | XP_015651490.1 | 5 - 6 | 1062 | 78 | ||
| XM_015796005.2 | XP_015651491.1 | 5 - 6 | 1062 | 78 | ||
| XM_015796007.2 | XP_015651493.1 | 5 - 6 | 1062 | 78 | ||
| XM_015796008.2 | XP_015651494.1 | 6 - 7 | 977 | 78 | ||
| XM_015796009.2 | XP_015651495.1 | 6 - 7 | 912 | 78 | ||
| GenBank mRNA | AK070222.1 | - | 5 - 6 | 1069 | 78 | |
| EU605826.1 | - | 4 - 5 | 749 | 78 | ||
More Information
Set Membership: |
|

LOC4347429
Genomic Map