Assay Details
Gene Symbol: LOC4347517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015755338.1 | XP_015610824.1 | 6 - 7 | 633 | 134 | |
GenBank mRNA | AK072867.1 | - | 6 - 7 | 629 | 134 | |
U78892.1 | - | 6 - 7 | 627 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() |
