Assay Details
| Gene Symbol: LOC4347517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755338.1 | XP_015610824.1 | 7 - 8 | 796 | 97 | |
| GenBank mRNA | AK072867.1 | - | 7 - 8 | 792 | 97 | |
| U78892.1 | - | 7 - 8 | 790 | 97 | ||
More Information
Set Membership: |
|

LOC4347517
Genomic Map