Assay Details
| Gene Symbol: LOC4348450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758643.2 | XP_015614129.1 | 29 - 30 | 7128 | 87 | |
| XM_015758644.2 | XP_015614130.1 | 29 - 30 | 7125 | 87 | ||
| GenBank mRNA | AK058817.1 | - | 1 - 2 | 686 | 87 | |
More Information
Set Membership: |
|

LOC4348450
Genomic Map