Assay Details
| Gene Symbol: LOC4348499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758903.2 | XP_015614389.1 | 1 - 2 | 190 | 80 | |
| GenBank mRNA | AK059907.1 | - | 1 - 2 | 149 | 80 | |
| AK102858.1 | - | 1 - 2 | 178 | 80 | ||
More Information
Set Membership: |
|

LOC4348499
Genomic Map