Assay Details
| Gene Symbol: LOC4348499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758903.2 | XP_015614389.1 | 3 - 4 | 358 | 62 | |
| GenBank mRNA | AK059907.1 | - | 3 - 4 | 317 | 62 | |
| AK102858.1 | - | 3 - 4 | 346 | 62 | ||
More Information
Set Membership: |
|

LOC4348499
Genomic Map