Assay Details
| Gene Symbol: LOC4348517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757316.2 | XP_015612802.1 | 2 - 3 | 403 | 72 | |
| GenBank mRNA | AK065782.1 | - | 2 - 3 | 404 | 72 | |
More Information
Set Membership: |
|

LOC4348517
Genomic Map