Assay Details
| Gene Symbol: LOC4348583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759124.1 | XP_015614610.1 | 1 - 2 | 219 | 108 | |
| GenBank mRNA | AK072672.1 | - | 1 - 2 | 61 | 108 | |
More Information
Set Membership: |
|

LOC4348583
Genomic Map