Assay Details
| Gene Symbol: LOC4348583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759124.1 | XP_015614610.1 | 5 - 6 | 522 | 151 | |
| GenBank mRNA | AK072672.1 | - | 5 - 6 | 364 | 151 | |
| JF969230.1 | - | 5 - 6 | 344 | 151 | ||
More Information
Set Membership: |
|

LOC4348583
Genomic Map