Assay Details
| Gene Symbol: LOC4324062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766684.2 | XP_015622170.1 | 10 - 11 | 1047 | 93 | |
| GenBank mRNA | AK100550.1 | - | 10 - 11 | 1032 | 93 | |
More Information
Set Membership: |
|

LOC4324062
Genomic Map