Assay Details
| Gene Symbol: LOC4324062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766684.2 | XP_015622170.1 | 11 - 12 | 1219 | 81 | |
| GenBank mRNA | AK100550.1 | - | 11 - 12 | 1204 | 81 | |
More Information
Set Membership: |
|

LOC4324062
Genomic Map