Assay Details
| Gene Symbol: LOC4324062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766684.2 | XP_015622170.1 | 4 - 5 | 450 | 75 | |
| GenBank mRNA | AK100550.1 | - | 4 - 5 | 435 | 75 | |
More Information
Set Membership: |
|

LOC4324062
Genomic Map