Assay Details
| Gene Symbol: LOC4324062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766684.2 | XP_015622170.1 | 6 - 7 | 639 | 66 | |
| GenBank mRNA | AK100550.1 | - | 6 - 7 | 624 | 66 | |
More Information
Set Membership: |
|

LOC4324062
Genomic Map