Assay Details
| Gene Symbol: LOC4325253 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766743.2 | XP_015622229.1 | 4 - 5 | 749 | 90 | |
| GenBank mRNA | AK061229.1 | - | 4 - 5 | 664 | 90 | |
More Information
Set Membership: |
|

LOC4325253
Genomic Map