Assay Details
| Gene Symbol: LOC4327178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766401.2 | XP_015621887.1 | 1 - 2 | 401 | 78 | |
| GenBank mRNA | AB255025.1 | - | 1 - 2 | 318 | 78 | |
| AK068523.1 | - | 1 - 2 | 405 | 78 | ||
More Information
Set Membership: |
|

LOC4327178
Genomic Map