Assay Details
| Gene Symbol: LOC4327178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766401.2 | XP_015621887.1 | 6 - 7 | 958 | 77 | |
| GenBank mRNA | AB255025.1 | - | 6 - 7 | 875 | 77 | |
| AK061638.1 | - | 3 - 4 | 345 | 77 | ||
| AK068523.1 | - | 6 - 7 | 962 | 77 | ||
More Information
Set Membership: |
|

LOC4327178
Genomic Map