Assay Details
| Gene Symbol: LOC4327178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766401.2 | XP_015621887.1 | 7 - 8 | 1097 | 127 | |
| GenBank mRNA | AB255025.1 | - | 7 - 8 | 1014 | 127 | |
| AK061638.1 | - | 4 - 5 | 484 | 127 | ||
| AK068523.1 | - | 7 - 8 | 1101 | 127 | ||
More Information
Set Membership: |
|

LOC4327178
Genomic Map