Assay Details
| Gene Symbol: LOC4327178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766401.2 | XP_015621887.1 | 9 - 10 | 1415 | 80 | |
| GenBank mRNA | AB255025.1 | - | 9 - 10 | 1332 | 80 | |
| AK061638.1 | - | 6 - 7 | 802 | 80 | ||
| AK068523.1 | - | 9 - 10 | 1419 | 80 | ||
More Information
Set Membership: |
|

LOC4327178
Genomic Map