Assay Details
| Gene Symbol: LOC4325777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765851.2 | XP_015621337.1 | 13 - 14 | 1494 | 79 | |
| GenBank mRNA | AK062011.1 | - | 9 - 10 | 950 | 79 | |
| AK070568.1 | - | 13 - 14 | 1463 | 79 | ||
| AK099208.1 | - | 13 - 14 | 1430 | 79 | ||
More Information
Set Membership: |
|

LOC4325777
Genomic Map