Assay Details
| Gene Symbol: LOC4325777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765851.2 | XP_015621337.1 | 15 - 16 | 1649 | 101 | |
| GenBank mRNA | AK062011.1 | - | 11 - 12 | 1105 | 101 | |
| AK070568.1 | - | 15 - 16 | 1618 | 101 | ||
| AK099208.1 | - | 15 - 16 | 1585 | 101 | ||
More Information
Set Membership: |
|

LOC4325777
Genomic Map