Assay Details
| Gene Symbol: LOC4325777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765851.2 | XP_015621337.1 | 16 - 17 | 1938 | 61 | |
| GenBank mRNA | AK062011.1 | - | 12 - 13 | 1394 | 61 | |
| AK070568.1 | - | 16 - 17 | 1907 | 61 | ||
| AK099208.1 | - | 16 - 17 | 1874 | 61 | ||
More Information
Set Membership: |
|

LOC4325777
Genomic Map