Assay Details
| Gene Symbol: LOC4325777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765851.2 | XP_015621337.1 | 5 - 6 | 572 | 128 | |
| GenBank mRNA | AK070568.1 | - | 5 - 6 | 541 | 128 | |
| AK099208.1 | - | 5 - 6 | 508 | 128 | ||
More Information
Set Membership: |
|

LOC4325777
Genomic Map